Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitroindole ≥98%

Supplier: Matrix Scientific
Synonyms: 5-Nitro-1H-indole

007298-25G
101792-386EA 58.46 USD InStock
101792-386
5-Nitroindole ≥98%
5-Nitroindole
Hazard code(s): Irritant, Irritant-Harmful, Store in Freezer


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0
Order Now
Specification Test Results
Melting point 141-142°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty