Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,5,6-Tetrafluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific
Warning
Synonyms: (2,3,5,6-Tetrafluorophenyl)methanol

007445-100G 007445-25G
101792-784EA 238.46 USD InStock
101792-784 101792-782
2,3,5,6-Tetrafluorobenzyl alcohol ≥98%
2,3,5,6-Tetrafluorobenzyl alcohol
Hazard code(s): Irritant


Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 79…80 °C (10mm)
Melting Pt: 32…34 °C
MDL Number: MFCD00792428
CAS Number: 4084-38-2
EINECS: 443-840-7
Order Now
Specification Test Results
Boiling point 79-80°C/10m
Melting point 32-34°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty