Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenethylamine ≥97%

Supplier: Matrix Scientific
Synonyms: 2-(4-Chlorophenyl)ethylamine

007540-25G 007540-5G
101792-998EA 41.54 USD InStock
101792-998 101793-000
4-Chlorophenethylamine ≥97%
4-Chlorophenethylamine
Hazard code(s): Irritant


Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 242…245 °C
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
Order Now
Specification Test Results
Boiling point 242°C-245°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty