Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzoic acid ≥98%

Supplier: Matrix Scientific
Synonyms: p-Iodobenzoic acid

008700-100G
101795-214EA 184.62 USD InStock
101795-214
4-Iodobenzoic acid ≥98%
4-Iodobenzoic acid
Hazard code(s): Irritant, Light Sensitive


Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: ∼265 °C (dec.)
Density: 2.184 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2
Order Now
Specification Test Results
Melting point ca 265°C (d

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty