Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromothiophene ≥98%

Supplier: Matrix Scientific

008719-25G 008719-100G
101795-280EA 44.62 USD InStock
101795-280 101795-282
2,3-Dibromothiophene ≥98%
2,3-Dibromothiophene
Hazard code(s): Irritant


Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 215…216 °C
Melting Pt: –17.5 °C
Density: 2.135 g/cm³
MDL Number: MFCD00005418
CAS Number: 3140-93-0
EINECS: 221-542-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Boiling point 215-216°C
Density 2.135
ND20 1.63

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty