Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrothiophenol ≥95%

Supplier: Matrix Scientific
Synonyms: 1-Mercapto-4-nitrobenzene4-Nitrobenzene-1-thiolp-Nitrothiophenol4-Nitrobenzenethiol

008768-100G 008768-25G
101795-408EA 583.08 USD InStock
101795-408 101795-410
4-Nitrothiophenol ≥95%
4-Nitrothiophenol
Hazard code(s): Irritant, Stench, Refrigerate


Formula: C₆H₅NO₂S
Melting Pt: 74…78 °C
MDL Number: MFCD00007343
CAS Number: 1849-36-1
EINECS: 217-436-4
Order Now
Specification Test Results
Melting point 74-78°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty