Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dichloropyridine ≥95%

Supplier: Matrix Scientific

011136-25G 011136-5G
101799-976EA 26.15 USD InStock
101799-976 101799-978
3,4-Dichloropyridine ≥95%
3,4-Dichloropyridine
Hazard code(s): Irritant, Store in Freezer


Formula: C₅H₃Cl₂N
Melting Pt: 22…23 °C
MDL Number: MFCD01861989
CAS Number: 55934-00-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty