Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloronicotinic acid ≥97%

Supplier: Matrix Scientific
Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid2,6-Dichloropyridine-3-carboxylic acid

011154-25G 011154-100G
101800-010EA 67.69 USD InStock
101800-010 101800-012
2,6-Dichloronicotinic acid ≥97%
2,6-Dichloronicotinic acid
Hazard code(s): Irritant


Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 143…146 °C
MDL Number: MFCD00075583
CAS Number: 38496-18-3
Order Now
Specification Test Results
Melting point 143-146°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty