Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxypicolinic acid ≥98%

Supplier: Matrix Scientific
Synonyms: 3-Hydroxypyridine-2-carboxylic acid

011218-5G 011218-25G
101800-148EA 69.23 USD InStock
101800-148 101800-150
3-Hydroxypicolinic acid ≥98%
3-Hydroxypicolinic acid
Hazard code(s): Irritant


Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 213…218 °C
MDL Number: MFCD00006294
CAS Number: 874-24-8
EINECS: 212-859-0
Order Now
Specification Test Results
Melting point 213-218°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty