Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-3-hydroxypyridine ≥99%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Bromo-3-pyridinol

011228-25G 011228-100G
101800-168EA 35.38 USD InStock
101800-168 102663-584
2-Bromo-3-hydroxypyridine ≥99%
2-Bromo-3-hydroxypyridine
Hazard code(s): Irritant


Formula: C₅H₄BrNO
MW: 174 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5
Order Now
Specification Test Results
Melting point 185-188°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty