Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoisonicotinic acid

Supplier: Matrix Scientific
Synonyms: 2-Iodopyridine-4-carboxylic acid

018244-1G
101805-714EA 198.46 USD InStock
101805-714
2-Iodoisonicotinic acid
2-Iodoisonicotinic acid
Hazard code(s): Irritant


Formula: C₆H₄INO₂
Melting Pt: 210…220 °C (dec.)
MDL Number: MFCD06800803
CAS Number: 58481-10-0
Order Now
Specification Test Results
Melting point 210-220°C(d

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty