Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-4-nitroaniline ≥97%

Supplier: Matrix Scientific
Warning

018278-25G
101805-802EA 46.15 USD InStock
101805-802
2-Fluoro-4-nitroaniline ≥97%
2-Fluoro-4-nitroaniline
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 122…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00034560
CAS Number: 369-35-7
EINECS: 206-719-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty