Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-3-nitropyridine ≥98%

Supplier: Matrix Scientific

018339-5G
101805-970EA 60 USD InStock
101805-970
2-Fluoro-3-nitropyridine ≥98%
2-Fluoro-3-nitropyridine
Hazard code(s): Toxic


Formula: C₅H₃FN₂O₂
MDL Number: MFCD03095068
CAS Number: 1480-87-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty