Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methylnicotinic acid ≥99%

Supplier: Matrix Scientific
Synonyms: 6-Methylpyridine-3-carboxylic acid6-Methyl-3-pyridinecarboxylic acid

021134-25G
101811-414EA 49.23 USD InStock
101811-414
6-Methylnicotinic acid ≥99%
6-Methylnicotinic acid
Hazard code(s): Irritant


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 214…215 °C
MDL Number: MFCD00006341
CAS Number: 3222-47-7
Order Now
Specification Test Results
Melting point 214-215°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty