Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromoanthranilic acid ≥98%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Amino-5-bromobenzoic acid

024631-100G 024631-25G
101818-500EA 21.54 USD InStock
101818-500 101818-502
5-Bromoanthranilic acid ≥98%
5-Bromoanthranilic acid
Hazard code(s): Irritant


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,01389
Order Now
Specification Test Results
Melting point 219-221°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty