Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-iodopyridine ≥98%

Supplier: Matrix Scientific
Warning

032363-5G
101833-574EA 36.92 USD InStock
101833-574
2-Bromo-5-iodopyridine ≥98%
2-Bromo-5-iodopyridine
Hazard code(s): Irritant


Formula: C₅H₃BrIN
MW: 283.89 g/mol
Melting Pt: 123…124 °C
MDL Number: MFCD03095201
CAS Number: 73290-22-9
Order Now
Specification Test Results
Melting point 123-124°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty