Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quinaldic acid ≥98%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Quinolinecarboxylic acidQuinoline-2-carboxylic acid

034842-100G
101838-480EA 113.85 USD InStock
101838-480
Quinaldic acid ≥98%
Quinaldic acid
Hazard code(s): Irritant


Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00006752
CAS Number: 93-10-7
EINECS: 202-218-3
Merck Index: 12,08227
Order Now
Specification Test Results
Melting point 156-159°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty