Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Ethylthio)acetic acid

Supplier: Matrix Scientific
Synonyms: (Ethylthio)aceticacid

036320-500MG
101841-126EA 146.15 USD InStock
101841-126
(Ethylthio)acetic acid
(Ethylthio)acetic acid


Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 164 °C (83 mmHg)
Melting Pt: –8.5 °C
Density: 1.1497 g/cm³ (20 °C)
MDL Number: MFCD00021769
CAS Number: 627-04-3
EINECS: 210-979-8
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty