Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloropurine ≥97%

Supplier: Matrix Scientific
Warning
Synonyms: 6-Chloro-9H-purine

037496-100G 037496-25G
102663-688EA 120 USD InStock
102663-688 101843-534
6-Chloropurine ≥97%
6-Chloropurine
Hazard code(s): Irritant


Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Boiling Pt: 185…192 °C (2 mmHg)
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty