Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-DL-Pro-OH

Supplier: Matrix Scientific
Synonyms: N-Boc-DL-proline1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

037561-25G
101843-666EA 126.15 USD InStock
101843-666
Boc-DL-Pro-OH
Boc-DL-Pro-OH
Hazard code(s): Irritant


Formula: C₁₀H₁₇NO₄
Melting Pt: 136 °C
MDL Number: MFCD00520915
CAS Number: 59433-50-0
Order Now
Specification Test Results
Melting point 136°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty