Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl isocyanate

Supplier: Matrix Scientific
Synonyms: 3-Isocyanatoprop-1-ene

037575-500MG
101843-692EA 290.77 USD InStock
101843-692
Allyl isocyanate
Allyl isocyanate
Hazard code(s): Flammable, Toxic


Formula: C₄H₅NO
MW: 83.09 g/mol
Boiling Pt: 87…89 °C
Density: 0.940 g/cm³
MDL Number: MFCD00002044
CAS Number: 1476-23-9
Order Now
Specification Test Results
Boiling point 87-89°C
Density 0.94
ND20 1.4185

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty