Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(4-(Dimethylamino)benzylidene)rhodanine

Supplier: Matrix Scientific
Synonyms: 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinonep-Dimethylaminobenzalrhodanine5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-onep-(Dimethylamino)benzal-5-rhodanine4-Dimethylaminobenzalrhodanine(5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

040155-500MG
101848-948EA 116.92 USD InStock
101848-948
5-(4-(Dimethylamino)benzylidene)rhodanine
5-(4-(Dimethylamino)benzylidene)rhodanine


Formula: C₁₂H₁₂N₂OS₂
MW: 264.37 g/mol
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty