Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobutan-1-ol ≥95%

Supplier: Matrix Scientific
Danger

041218-25G 041218-5G
101850-954EA 189.23 USD InStock
101850-954 101850-952
4-Aminobutan-1-ol ≥95%
4-Aminobutan-1-ol
Hazard code(s): Corrosive, Air Sensitive, Hygroscopic


Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206 °C
Melting Pt: 16…18 °C
Density: 0.967 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008230
CAS Number: 13325-10-5
EINECS: 236-364-4
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Boiling point 206°C
Melting point 16-18°C
Density 0.967
ND20 1.461

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty