Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Alanine

Supplier: Matrix Scientific
Synonyms: D(-)-Alanineα-Alanine(R)-(-)-AlanineAlanineD-α-AlanineH-D-Ala-OH

041261-100G 041261-25G
101851-094EA 60 USD InStock
101851-094 101851-092
D-Alanine
D-Alanine
Hazard code(s): Irritant


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: ∼295 °C (dec.)
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty