Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Arginine

Supplier: Matrix Scientific
Warning
Synonyms: (R)-2-Amino-5-guanidinopentanoic acidH-D-Arg-OH(R)-(-)-Arginine

041283-25G
101851-192EA 106.15 USD InStock
101851-192
D-(-)-Arginine
D(-)-Arginine


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00063116
CAS Number: 157-06-2
EINECS: 205-866-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty