Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichloro-5-methoxyaniline ≥95%

Supplier: Matrix Scientific

044492-5G 044492-25G
101866-154EA 55.38 USD InStock
101866-154 101866-156
2,4-Dichloro-5-methoxyaniline ≥95%
2,4-Dichloro-5-methoxyaniline
Hazard code(s): Irritant


Formula: C₇H₇Cl₂NO
Melting Pt: 49…51 °C
MDL Number: MFCD00974410
CAS Number: 98446-49-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 49-51°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty