Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

045699-5G
101872-172EA 35.38 USD InStock
101872-172
4-Amino-2-fluorobenzoic acid ≥95%
4-Amino-2-fluorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 216…219 °C
MDL Number: MFCD01569397
CAS Number: 446-31-1
Order Now
Specification Test Results
Melting point 216-219°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty