Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraphenylmethane ≥96%

Supplier: Matrix Scientific

054033-5G 054033-1G
101905-904EA 106.15 USD InStock
101905-904 101905-906
Tetraphenylmethane ≥96%
Tetraphenylmethane
Hazard code(s): Irritant


Formula: C₂₅H₂₀
MW: 320.43 g/mol
Boiling Pt: 431 °C
Melting Pt: 280…285 °C
Density: 1.217 g/cm³ (20 °C)
MDL Number: MFCD00014428
CAS Number: 630-76-2
EINECS: 211-144-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/63/2299863.jpg
Order Now
Specification Test Results
Boiling point 431°C
Melting point 280-285°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty