Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-4-iodotoulene ≥98%

Supplier: Matrix Scientific
Danger
Synonyms: 1-(Bromomethyl)-4-iodobenzene4-Iodobenzyl bromideα-Bromo-p-iodotoulene

056743-5G
101911-310EA 109.23 USD InStock
101911-310
α-Bromo-4-iodotoulene ≥98%
α-Bromo-4-iodotoulene
Hazard code(s): Corrosive, Lachrymator, Light Sensitive, Moisture Sensitive, Refrigerate, Stored under Argon


Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 115…120 °C (0.6 mmHg)
Melting Pt: 77…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Melting point 79-81°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty