Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anthranilamide ≥95%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Aminobenzamide

056883-250G 056883-100G
102833-196EA 26.15 USD InStock
102833-196 101911-590
Anthranilamide ≥95%
Anthranilamide
Hazard code(s): Irritant


Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 108…110 °C
Flash Pt: >185 °C
Storage Temperature: Ambient
MDL Number: MFCD00007981
CAS Number: 88-68-6
EINECS: 201-851-2
Order Now
Specification Test Results
Melting point 108-110°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty