Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α-Dimethoxytoluene

Supplier: Matrix Scientific
Warning
Synonyms: Benzaldehyde dimethyl acetal

056890-100G 056890-500G
101911-610EA 227.69 USD InStock
101911-610 101911-608
α,α-Dimethoxytoluene
α,α-Dimethoxytoluene
Hazard code(s): Irritant


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 87…89 °C (18mm)
Density: 1.01 g/cm³
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0
Order Now
Specification Test Results
Boiling point 87-89°C/18m
Density 1.01

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty