Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Hydroxyphenyl)acetonitrile

Supplier: Matrix Scientific
Synonyms: 4-Hydroxybenzyl cyanide2-(4-Hydroxyphenyl)acetonitrile

057097-25G
101912-360EA 29.23 USD InStock
101912-360
(4-Hydroxyphenyl)acetonitrile
(4-Hydroxyphenyl)acetonitrile
Hazard code(s): Irritant


Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 69…71 °C
MDL Number: MFCD00002383
CAS Number: 14191-95-8
EINECS: 238-046-0
Order Now
Specification Test Results
Melting point 69-71°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty