Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylphenol

Supplier: Matrix Scientific
Danger
Synonyms: 4-Amylphenol

057169-25G
101912-618EA 284.62 USD InStock
101912-618
4-Pentylphenol
4-Pentylphenol
Hazard code(s): Irritant


Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 24 °C
Density: 0.966 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00020211
CAS Number: 14938-35-3
EINECS: 239-015-4
UN: 2430
ADR: 8,III
Order Now
Specification Test Results
Melting point 24°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty