Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2-chlorobenzoic acid ≥95%

Supplier: Matrix Scientific
Warning

057675-100G
101913-526EA 70.77 USD InStock
101913-526
5-Amino-2-chlorobenzoic acid ≥95%
5-Amino-2-chlorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.519 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007845
CAS Number: 89-54-3
EINECS: 201-916-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty