Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citraconic acid

Supplier: Matrix Scientific
Synonyms: 2-Methyl-2-butenedioic acid

058308-25G
101914-558EA 69.23 USD InStock
101914-558
Citraconic acid
Citraconic acid


Formula: C₅H₆O₄
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 98…99 °C
Density: 1.617 g/cm³ (20 °C)
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty