Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Diphenylcarbazide

Supplier: Matrix Scientific
Warning
Synonyms: sym. Diphenylcarbazidesym-DiphenylcarbazideN'',N'''-Diphenylcarbonohydrazide1,5-DiphenylcarbohydrazideDiphenylcarbazide1,5-Diphenylcarbonohydrazide

058311-500MG
101914-560EA 306.15 USD InStock
101914-560
1,5-Diphenylcarbazide
1,5-Diphenylcarbazide


Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 170…172 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://us-legacy.vwr.com/stibo/web/std.lang.all/46/13/2374613.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty