Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrachloro-p-benzoquinone

Supplier: Matrix Scientific
Warning
Synonyms: Tetrachloroquinonep-Chloranil2,3,5,6-Tetrachlorobenzo-1,4-quinone2,3,5,6-Tetrachloro-1,4-benzoquinoneTetrachloro-1,4-benzoquinone4-Chloranil

058465-25G 058465-100G
101914-864EA 84.62 USD InStock
101914-864 101914-862
Tetrachloro-p-benzoquinone
Tetrachloro-p-benzoquinone
Hazard code(s): Irritant-Harmful, Light Sensitive


Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 287…290 °C
Density: 1.97 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
Merck Index: 13,02088
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty