Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trimethylsiloxy)cyclohexene ≥98%

Supplier: Matrix Scientific
Synonyms: (Cyclohex-1-en-1-yloxy)trimethylsilane

059620-25G
101918-660EA 118.46 USD InStock
101918-660
1-(Trimethylsiloxy)cyclohexene ≥98%
1-(Trimethylsiloxy)cyclohexene
Hazard code(s): Flammable, Moisture Sensitive, Refrigerate


Formula: C₉H₁₈OSi
MW: 170.33 g/mol
Boiling Pt: 64…65 °C (15mm)
MDL Number: MFCD00001541
CAS Number: 6651-36-1
EINECS: 229-675-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty