Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Butoxybenzoyl chloride

Supplier: Matrix Scientific
Synonyms: 4-N-butoxybenzoylchloride

061815-500MG
101922-722EA 364.62 USD InStock
101922-722
4-Butoxybenzoyl chloride
4-Butoxybenzoyl chloride


Formula: C₁₁H₁₃ClO₂
MW: 212.68 g/mol
Boiling Pt: 191…193 °C (33 mmHg)
Density: 1.121 g/cm³ (20 °C)
MDL Number: MFCD00000689
CAS Number: 33863-86-4
EINECS: 251-703-6
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty