Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propiolaldehyde diethyl acetal

Supplier: Matrix Scientific
Synonyms: 3,3-Diethoxy-1-propynePropioaldehyde diethyl acetal3,3-Diethoxypropyne3,3-Diethoxyprop-1-ynePropargylaldehyde diethyl acetalPropynal diethyl acetal

064612-25G 064612-5G
101928-032EA 89.23 USD InStock
101928-032 101928-034
Propiolaldehyde diethyl acetal
Propiolaldehyde diethyl acetal
Hazard code(s): Flammable


Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 138…139.5 °C
Density: 0.894 g/cm³
MDL Number: MFCD00009237
CAS Number: 10160-87-9
EINECS: 233-430-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty