Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trimethylsilyl)thiazole ≥98%

Supplier: Matrix Scientific

069609-25G 069609-1G 069609-5G
101941-062EA 87.69 USD InStock
101941-062 101941-066 101941-064
2-(Trimethylsilyl)thiazole ≥98%
2-(Trimethylsilyl)thiazole
Hazard code(s): Irritant


Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
MDL Number: MFCD00066274
CAS Number: 79265-30-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty