Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dichloroacetone ≥96%

Supplier: Matrix Scientific

070074-500G 070074-100G
102833-470EA 373.85 USD InStock
102833-470 101943-202
1,3-Dichloroacetone ≥96%
1,3-Dichloroacetone
Hazard code(s): Toxic


Formula: ClH₂CC(O)CH₂Cl
MW: 126.97 g/mol
Boiling Pt: 172…173 °C
Melting Pt: 40…44 °C
Density: 1.383 g/cm³
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000937
CAS Number: 534-07-6
EINECS: 208-585-6
UN: 2649
ADR: 6.1,II
Merck Index: 13,03077
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty