Would you like to log in again?
Axygen® 384-Well PCR Plates, Corning
Supplier: Corning
Axygen®
PCR-384-LC480-W
89131-748CS 1198.81 USD InStock
89131-748
Axygen® 384-Well PCR Plates, Corning
Microplates PCR Plates
Axygen's plates are made of 99.9% pure virgin polypropylene and specifically designed for PCR applications
The Axygen brand microplates for the Roche Lightcycler® 480 Real-Time PCR system are a perfect to maximize the potential of the LightCycler 480. The consistent thin wall design presents optimized assay conditions, while plates are manufactured from white polypropylene to magnify signal intensity and prevent “cross-talk” between wells. Printed black alphanumerics make it easier to locate wells and orient the plate. All plates are free of any detectable Rnase/Dnase and endotoxins (pyrogens).
Plates are ideal for thermal transfer applications because their ultra-thin walls accelerate heat transfer to provide shorter cycles. Certified RNase-, DNase-, and pyrogen-free to ensure purity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty