Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4,4,4-trifluorobutyric acid ≥97%

Supplier: Matrix Scientific
Synonyms: (±)-2-Amino-4,4,4-trifluorobutyric acid

001212-1G 001212-250MG
101781-162EA 84.62 USD InStock
101781-162 101781-164
2-Amino-4,4,4-trifluorobutyric acid ≥97%
(±)-2-Amino-4,4,4-trifluorobutyric acid
Hazard: Irritant


Formula: C₄H₆F₃NO₂
MW: 157.09 g/mol
Melting Pt: 260…265 °C
MDL Number: MFCD00041417
CAS Number: 15959-93-0
Order Now
Specification Test Results
Melting point 260-265°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty