Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-DL-glutamic acid ≥97%

Supplier: Matrix Scientific

002819-1G
101784-712EA 1069.23 USD InStock
101784-712
4-Fluoro-DL-glutamic acid ≥97%
4-Fluoro-DL-glutamic acid
Hazard: Irritant


Formula: C₅H₈FNO₄
Melting Pt: 185 °C (dec.)
MDL Number: MFCD00055778
CAS Number: 2708-77-2
Order Now
Specification Test Results
Melting point 185°C(dec)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty