Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Imidazo[1,2-a]pyrid-2-yl)aniline ≥95%

Supplier: Matrix Scientific

044596-500MG 044596-1G
101866-642EA 921.54 USD InStock
101866-642 101866-644
4-(Imidazo[1,2-a]pyrid-2-yl)aniline ≥95%
4-(Imidazo[1,2-a]pyrid-2-yl)aniline
Hazard: Irritant


Formula: C₁₃H₁₁N₃
MW: 209.252 g/mol
Melting Pt: 204…206 °C
MDL Number: MFCD00107394
CAS Number: 139705-74-1
Order Now
Specification Test Results
Melting point 204-206°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty