Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-3-Fluoropyrrolidine hydrochloride ≥98%, ee 98%

Supplier: Matrix Scientific
Synonyms: (S)-3-Fluoropyrrolidine hydrochloride

009062-1G 009062-500MG
101796-244EA 75.38 USD InStock
101796-244 101796-242
(S)-(+)-3-Fluoropyrrolidine hydrochloride ≥98%, ee 98%
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Hazard: Irritant/Store in Freezer


Formula: C₄H₉ClFN
MW: 125.57 g/mol
Melting Pt: 182…185 °C
MDL Number: MFCD04038718
CAS Number: 136725-53-6
Order Now
Specification Test Results
Melting point 182-185°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty