Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoroacetyl)benzoic acid ≥98%

Supplier: Matrix Scientific
Synonyms: 4-(2,2,2-trifluoroacetyl)benzoic acid

005141-1G
101788-318EA 87.69 USD InStock
101788-318
4-(Trifluoroacetyl)benzoic acid ≥98%
4-(Trifluoroacetyl)benzoic acid
Hazard: Irritant


Formula: C₉H₅F₃O₃
MW: 218.13 g/mol
Melting Pt: 177…179 °C
MDL Number: MFCD00052340
CAS Number: 58808-59-6
Order Now
Specification Test Results
Melting point 177-179°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty