Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Thiophenedicarboxylic acid ≥97%

Supplier: Matrix Scientific
Synonyms: Thiophene-2,5-dicarboxylic acid

020591-5G
101810-382EA 32.31 USD InStock
101810-382
2,5-Thiophenedicarboxylic acid ≥97%
2,5-Thiophenedicarboxylic acid
Hazard: Irritant


Formula: C₆H₄O₄S
MW: 172.16 g/mol
Melting Pt: 355…360 °C
MDL Number: MFCD00016896
CAS Number: 4282-31-9
Order Now
Specification Test Results
Melting point 355-360°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty