Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal ≥95%

Supplier: Matrix Scientific
Synonyms: 8-Amino-1,4-dioxaspiro[4.5]decane-8-carboxylic Acid

024829-250MG 024829-1G
101819-008EA 73.85 USD InStock
101819-008 101819-010
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal ≥95%
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal
Hazard: Irritant/Store in Freezer


Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 301…304 °C
MDL Number: MFCD03790906
CAS Number: 54621-18-0
Order Now
Specification Test Results
Melting point 301-304°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty